Learn More
Cytiva PD SpinTrap™ G-25 Desalting Columns
Use for convenient and fast small-scale cleanup of proteins and other large biomolecules (>5000 Mr).  Cytiva Life Sciences™ PD SpinTrap™ G-25 Desalting Columns provide the option of running multiple samples in parallel.
Brand:  Cytiva 28-9180-04
306.85 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Additional Details : Weight : 0.18450kg
Product Code. 11783329
  • 323.00 EUR / Pack of 50
Estimated Shipment: 31-05-2024
Description
  • High recovery (70 to 90%, biomolecule dependent) for a range of applications, such as desalting, buffer exchange, and removal of low-molecular weight compounds
  • High desalting capacity (>85%); prepacked with 500μL Sephadex™ G-25 Medium
  • Reproducible, high throughput, parallel cleanup of protein samples
  • Sample preparation in the range of 100 to 180 μL without sample dilution
  • For use with standard microcentrifuge
Specifications
Specifications
Range of applications such as desalting, buffer exchange, and removal of low-molecular weight compounds
≈600 μL
50 Pk.