Learn More
GeneBLAzer™ P2RY4-NFAT-bla CHO-K1 Cells
GeneBLAzer™ P2RY4-NFAT-bla CHO-K1 Cells contain the human Purinergic Receptor P2Y4.
Brand:  GeneBLAzer™ K1863
Additional Details : Weight : 0.01000kg
Product Code. 10535854
  • 15150.00 EUR / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
  • Accession Number: NM_002565.3
  • Population Status: Pool of Stable Cells
  • GeneBLAzer™ P2RY4-NFAT-bla CHO-K1 cells contain the human Purinergic Receptor P2Y4 (P2RY4), stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat.no.K1078) contain a beta-lactamase reporter gene under control of the NFAT Response Element. The GeneBLAzer™ P2RY4-NFAT-bla CHO-K1 cells are functionally validated for Z’-factor and EC50 concentrations of UTP. In addition, GeneBLAzer™P2RY4-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.
Specifications
Specifications
Liquid Nitrogen
GPCRs
Dividing Cells
Fluorescent
1 Vial