Learn More
Merck Chemicals ORBO™ 43 Supelpak-20 specially treated Amberlite XAD-2 (20/40), 100/50 mg W,W,W (specially treated) separators
Used for gas and vapor sampling
Brand: Merck Chemicals 20258
Additional Details : Weight : 3.00000kg
Description
ORBO™ sorbent tubes contain two beds of the same selective adsorbent separated by glass wool or foam, for gas and vapor sampling. The dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.Specifications
Bed A 100 mg/Bed B 50 mg | |
20 to 40 Mesh | |
100 mm |
W,W,W (Specially Treated) Separators | |
50/Pk. | |
8 mm |
ORBO is a trademark of Sigma-Aldrich Co. LLC