missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOP14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | NOP14 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16845195
|
Proteintech
26854-1-AP-20UL |
20 μL |
140.00 EUR
20µL |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16835195
|
Proteintech
26854-1-AP-150UL |
150 μL |
399.00 EUR
150µL |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Involved in nucleolar processing of pre-18S ribosomal RNA. Has a role in the nuclear export of 40S pre-ribosomal subunit to the cytoplasm.Specifications
NOP14 | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P78316, Q8R3N1 | |
289724, 75416, 8602 | |
NOP14 Fusion Protein Ag25476 | |
Primary | |
-20°C | |
NOP14 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C4orf9, NOL14, NOP14, Nucleolar complex protein 14, Nucleolar protein 14, RES4 25, RES425 | |
Nop14 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |