Learn More
Fisherbrand™ Nickel Chattaway Spatulas
Scrapes and lifts materials from bottles or flasks. Fisherbrand™ Nickel Chattaway Spatulas are non-magnetic with a bright finish.
Brand:  Fisherbrand™
Additional Details : Weight : 0.08280kg
 View more versions of this product
Product Code. 11583462
  • 58.60 EUR / Pack of 5
Estimated Shipment: 17-06-2024
Description
  • Manufactured from Nickel
  • Bent and rounded end scrapes and lifts materials from bottles and flasks
Specifications
Specifications
5 Pk.
10 mm
200 x 10 mm
200 mm
Chattaway Spatula
3.6 mm
Nickel