missing translation for 'onlineSavingsMsg'
Learn More

Nanog Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA585110

Additional Details : Weight : 0.01000kg

Product Code. 16372055

  • 474.00 EUR / 100µL
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Keep as concentrated solution. Predicted reactivity: Chimpanzee (98%). Positive Control: HeLa nucleus, NIH-3T3, mouse ESC. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

NANOG (Nanog homeobox) is a divergent homeodomain protein that directs pluripotency and differentiation of undifferentiated embryonic stem cells. NANOG mRNA is present in pluripotent mouse and human cell lines, and absent from differentiated cells. Human NANOG protein shares 52% overall amino acid identity with the mouse protein, and 85% identity in the homeodomain. Human NANOG maps to gene locus 12p13.31, while the mouse NANOG maps to gene loci 6 F2. Murine embryonic NANOG expression is detected in the inner cell mass of the blastocyst. Research studies have shown that high levels of human NANOG expression in the undifferentiated N-Tera embryonal carcinoma cell line. Further, NANOG is a transcription regulator involved in inner cell mass and embryonic stem (ES) cells proliferation and self-renewal. The role of NANOG in embryonic development suggested that it might be useful in the creation of stem cells that might be useful in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing POU5F1 (also known as Oct-4), another germline-specific transcription factor, and the transcription factors Sox2, Klf4 and Lin28 along with c-Myc in mouse fibroblasts. Experiments have demonstrated that iPS cells could be generated using expression plasmids expressing NANOG, Sox2, KlfF4 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine. When overexpressed, NANOG promotes cells to enter into S phase and proliferation. Diseases associated with dysfunction in the NANOG protein include tetracarcinoma and germ cell/embryonal cancer.
Specifications

Specifications

Nanog
Polyclonal
Unconjugated
NANOG
2410002E02Rik, early embryo specific expression NK family, early embryo specific expression NK-type homeobox protein, Ecat4, ENK, ES cell-associated protein 4, FLJ12581, HGNC:20857, hNanog, Homeobox protein NANOG, homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog, Nanog homeobox
Rabbit
Antigen affinity chromatography
RUO
71950, 79923
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
0.5 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal, pH 7
Q80Z64, Q9H9S0
NANOG
Recombinant protein encompassing a sequence within the center region of human NANOG.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers