Learn More
Eppendorf™ MTP Bucket Retrofit Kit
Designed for use with Eppendorf Rotor A-4-62-MTP
Brand: Eppendorf™ 5810702003
1211.00 EUR valid until 2024-12-31
Use promo code "23292" to get your promotional price.
Additional Details : Weight : 3.15000kg
Description
- Microtiter plate bucket
- For swing bucket rotor 4 x 250mL
- For 4 microtitre plates or 1 deepwell plate
Specifications
MTP Bucket Retrofit Kit | |
Rotor A-4-62-MTP |