Learn More
Laminarin
Product Code. p-7043204
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15417459
Brand: Thermo Scientific Alfa Aesar J66193.MD
Request Bulk or Custom Format
CAS: 9008-22-4 | C18H32O16 | 504.438 g/mol
Laminarin is a storage glucan and is now used to study it’s interaction with laminarinase as its substrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Laminarin is a storage glucan and is now used to study it’s interaction with laminarinase as its substrate.
Solubility
Soluble in dimethyl sulfoxide (slightly, heated). Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9008-22-4
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00046691
InChI Key DBTMGCOVALSLOR-DEVYUCJPSA-N
Synonym laminarin, laminaran
PubChem CID 46173707
IUPAC Name (2S,3R,4S,5R,6R)-4-[(2S,3R,4S,5R,6R)-3,5-dihydroxy-6-(hydroxymethyl)-4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,5-triol
SMILES C(C1C(C(C(C(O1)O)O)OC2C(C(C(C(O2)CO)O)OC3C(C(C(C(O3)CO)O)O)O)O)O)O
Specifications
Color White
Quantity 250 mg
Merck Index 14,5350
Solubility Information Soluble in dimethyl sulfoxide (slightly,heated). Slightly soluble in water.
Optical Rotation −45° (c=2 in Water)
Physical Form Powder
Chemical Name or Material Laminarin