missing translation for 'onlineSavingsMsg'
Learn More

KV1.3 (KCNA3) (extracellular) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA577618

Additional Details : Weight : 0.01000kg

Product Code. 15924125

  • 596.00 EUR / 50µL
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.

In neurons voltage-dependent potassium channels are key determits of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
Specifications

Specifications

KV1.3 (KCNA3) (extracellular)
Polyclonal
Unconjugated
KCNA3
KCNA3
Rabbit
Antigen affinity chromatography
RUO
16491, 29731, 3738
-20°C
Lyophilized
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
0.8 mg/mL
PBS with 1% BSA and 0.05% sodium azide; pH 7.4
P15384, P16390, P22001
Kcna3
Peptide KDYPASTSQDSFEA(C), corresponding to amino acid residues 263-276 of human Kv 1.3.
50 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers