Learn More
Hanna Instruments™ K-Type Waterproof Thermocouple Thermometers
A digital, waterproof thermometer that offers high accuracy temperature measurements using a wide variety of interchangeable probes.
Brand:  Hanna Instruments™ HI935005
Additional Details : Weight : 0.47700kg
Product Code. 10076151
  • 204.00 EUR / Each
Estimated Shipment: 29-05-2024
Description
  • K-Type Thermocouple Thermometer is a digital, waterproof thermometer that can be used with a wide variety of K-type probes
  • Offers high accuracy temperature measurements using a wide variety of interchangeable probes
  • Offers two measurement ranges from -50.0 to 199.9°C and 200 to 1350°C which can also be displayed in °F (-58.0 to 399.9°F and 400 to 2462°F)
  • With a ±0.2% full scale accuracy, is perfectly suited for temperature measurements in the laboratory or the field