Learn More
Lonza™ Rat Neonatal Dorsal Root Ganglion (DRG) Neurons
Click to view available options
Media Recommendation:
PNGM™ BulletKit™ Medium
Unit Size:
Each
Description
Cryopreserved Rat DRG cells thawed, cultured and immunostained with anti-Peripherin(green) and antiTuj-3 (red).
Specifications
Specifications
| Content And Storage | Cryopreserved |
| Cell Type | Neural Cells |
| Form | Cryopreserved |
| Species | Rat |
| Media Recommendation | PNGM™ BulletKit™ Medium |
| Product Type | Rat Dorsal Root Ganglion Neurons |
| Source | Rat Dorsal Root Ganglion Neurons |