Learn More
Deutsch Neumann™ Grey Natural Rubber Filter Ring with Flange
Brand:  Deutsch Neumann™ 2350031
Additional Details : Weight : 0.80000kg
Product Code. 11966116
  • 1.50 EUR / Each
Estimated Shipment: 05-06-2024
Specifications
Gray
1Each
56 mm
Natural Rubber
1 Each
20 mm