Learn More
TAP1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of TAP1.
Brand: Abnova PAB27600.100ug
Additional Details : Weight : 0.02000kg
Description
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is involved in the pumping of degraded cytosolic peptides across the endoplasmic reticulum into the membrane-bound compartment where class I molecules assemble. Mutations in this gene may be associated with ankylosing spondylitis, insulin-dependent diabetes mellitus, and celiac disease. [provided by RefSeq
Sequence: QKFREKLQEIKTSpecifications
TAP1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (1-3ug/ml) The optimal working dilution should be determined by the end user. | |
TAP1 | |
TAP1 | |
A synthetic peptide corresponding to internal region of human TAP1. | |
100 μg | |
Primary | |
Human |
Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of TAP1. | |
In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
ABC17/ABCB2/APT1/D6S114E/FLJ26666/FLJ41500/PSF1/RING4/TAP1*0102N/TAP1N | |
Goat | |
Affinity Purified | |
RUO | |
6890 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only