missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STX6, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of STX6.
Brand: Abnova PAB7011.100ug
97.82 EUR valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Additional Details : Weight : 0.02000kg
Description
Sequence: C-QALAERKNRQASpecifications
STX6 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
STX6 | |
Goat | |
Antigen affinity purification | |
RUO | |
10228 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of STX6. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
STX6 | |
A synthetic peptide corresponding to human STX6. | |
100 μg | |
Primary | |
Human | |
Liquid |
For Research Use Only