missing translation for 'onlineSavingsMsg'
Learn More

Geranyl acetate, 98%

Product Code. p-7024704
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458697 25 g 25g
11468697 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458697 Supplier Thermo Scientific Alfa Aesar Supplier No. A19864.14

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 105-87-3 | C12H20O2 | 196.29 g/mol

It is used primarily as a component of perfumes for creams and soaps and as a flavoring ingredient. It is used particularly in rose, lavender and geranium formulations where a sweet fruity or citrus aroma is desired. It is found to have has anti-oxidant, anti-inflammatory and anti-proliferative activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used primarily as a component of perfumes for creams and soaps and as a flavoring ingredient. It is used particularly in rose, lavender and geranium formulations where a sweet fruity or citrus aroma is desired. It is found to have has anti-oxidant, anti-inflammatory and anti-proliferative activities.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 105-87-3
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00015037
InChI Key HIGQPQRQIQDZMP-DHZHZOJOSA-N
Synonym geranyl acetate, geraniol acetate, bay pine oyster oil, acetic acid, geraniol ester, trans-3,7-dimethyl-2,6-octadien-1-yl acetate, geranyl ethanoate, acetic acid, geranyl ester, unii-3w81yg7p9r, geranyl acetate, cis, 3,7-dimethyl-2,6-octadienyl acetate
PubChem CID 1549026
ChEBI CHEBI:5331
IUPAC Name [(2E)-3,7-dimethylocta-2,6-dienyl] acetate
SMILES CC(C)=CCC\C(C)=C\COC(C)=O

Spezifikation

Density 0.91
Boiling Point 137°C to 139°C (25 mmHg)
Flash Point 104°C (219°F)
Assay Percent Range 0.98
Odor Pleasant
Refractive Index 1.461
Quantity 25 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 196.29
Percent Purity 98%
Chemical Name or Material Geranyl acetate
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Geranyl acetate
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Sicherheitshinweis
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Geranyl acetate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.