missing translation for 'onlineSavingsMsg'
Learn More

FCGRT Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA597738

Additional Details : Weight : 0.01000kg

Product Code. 16500572

  • 422.00 EUR / 100µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FCGRT binds to the Fc region of monomeric immunoglobulins gamma. It mediates the uptake of IgG from milk. It plays a possible role in transfer of immunoglobulin G from mother to fetus.
Specifications

Specifications

FCGRT
Polyclonal
Unconjugated
FCGRT
alpha-chain, Fc fragment of IgG receptor and transporter, Fc fragment of IgG, receptor, transporter, alpha, Fc receptor, IgG, alpha chain transporter, FCGRT, FCRN, FcRn alpha chain, heavy chain of the major histocompatibility complex class I-like Fc receptor, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, immunoglobulin receptor, intestinal, heavy chain, major histocompatibility complex class I-like Fc receptor, neonatal Fc receptor, neonatal Fc-receptor for Ig, transmembrane alpha chain of the neonatal receptor
Rabbit
Protein G
RUO
14132, 2217
-20°C or -80°C if preferred
Liquid
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1.2 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300, pH 7.4
P55899, Q61559
FCGRT
Recombinant Human IgG receptor FcRn large subunit p51 protein (24-297AA)
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers