Learn More
Falcon™ 96-Well, Non-Treated, V-Shaped-Bottom Microplate
Color:Clear; Sterility:Non-sterile
Brand:  Falcon™ 353263
Additional Details : Weight : 4.05000kg
Product Code. 10688892
  • 325.47 EUR / Pack of 100
Estimated Shipment: 05-06-2024
Description
Falcon™ 96-well Polypropylene Microplate, not treated, V-bottom, 25/pack, 50/case : The Falcon 96 well Polypropylene Library Storage Plate is convenient for compound storage : Notch A1-H1
  • Exceedingly rigid and flat plate for consistent automated handling
  • Plate withstands repeated freeze-thaws down to -20°C
  • Low retention polypropylene and round-well design give low residual volumes after pipetting (<0.1 μL)
  • Raised well edges and flatness for heat or adhesive sealing
  • In addition to the round-bottom Falcon 96 well Library Storage Plate, V-bottom formats are also available.
  • Specifications
    Specifications
    Standard
    Not Treated
    Polypropylene
    Non-sterile
    Conical Bottom (V-well)
    25/Pk.
    8 x 12
    Clear
    96
    Non-treated
    100/Cs.
    100/Cs.