Learn More
Lonza™ EGM-MV Endotheial Medium SingleQuot Kit
Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. EGM-MV Endothelial Medium SingleQuot Kit is part of a collection of tools optimized to grow certain cell types.
Brand:  Lonza™ CC-4143
Additional Details : Weight : 0.00050kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Product Code. 11625500
  • 359.00 EUR / Each
Estimated Shipment: 29-07-2024
Description
Media products are supplied in several formats:
  • Basal Medium -– contains no growth factors, cytokines, or supplements
  • SingleQuots™ Kit – growth factors, cytokines, and supplements
  • BulletKit™ – basal medium and SingleQuots™ Kit packaged together
The following Endothelial Cell Growth Medium BulletKits™ are available:
  • EGM™-2 BulletKit™ – Optimized for certain HUVECs, Aortic and Pulmonary artery endothelial cells. Contains 2% FBS and VEGF for rapid proliferation. Does not contain BBE.
  • EGM™ BulletKit™ – Optimized for HUVECs cat. no. CC-2517 and CC-2519, contains 2% FBS and BBE. Does not contain exogenous VEGF.
  • New! EGM™-Plus BulletKit™ – Enhanced version of EGM™ Medium. Optimized for HUVECs cat. no. CC-2935. Contains 2% FBS and BBE. Does not contain exogenous VEGF. Faster proliferation than HUVECs isolated in EGM™ Medium.
  • EGM™-2MV BulletKit™ – Coronary and Iliac artery, human microvascular endothelial cells, contains 5% FBS and VEGF for rapid proliferation. Does not contain BBE.
  • EGM™ -MV BulletKit™ – Bovine Microvascular Cells, contains 5% FBS and BBE. Does not contain exogenous VEGF.
Specifications
Specifications
1 kit