Learn More
Jiangsu HanHeng Medical Technology Disposable Sampling Swab
Standard nasopharyngeal swab with 100 mm breakpoint
Brand: Jiangsu HanHeng Medical Technology R12544
336.30 EUR valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Additional Details : Weight : 0.11340kg
Description
- Standard nasopharyngeal swab with 100 mm breakpoint with plastic handle
Specifications
Nasopharyngeal | |
CE | |
Plastic | |
100 mm |
Plastic | |
Nylon | |
36 Months |