Learn More
Invitrogen™ Custom Primer, 100% QC Illumina Each
Custom Primer, 100% QC Illumina Each
Brand:  Invitrogen™ 10668119
Additional Details : Weight : 0.00500kg
Product Code. 10543433
  • 0.53 EUR / Each
Estimated Shipment: 14-05-2024
to see stock.
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
Specifications
100% QC Illumina Primer, 25 nmol
100% QC Illumina Primer
Room Temperature
25 nmol