Learn More
Fisherbrand™ Mixing Cup for Tube Mixing
With an easy to clean design allowing the whole platform to be autoclaved, this mixing cup for tube mixing is ideal for laboratory, research, and other environments.
Brand:  Fisherbrand™
Additional Details : Weight : 0.05640kg
Product Code. 10489193
  • 42.10 EUR / Each
Estimated Shipment: 01-07-2024
Description
  • Mixing platform for CM-2 Cyclone+
  • Easy push on and lift off design
  • Easy to clean design allowing whole platform to be autoclaved
Specifications
Specifications
Cup
For Use with Whirlimixer Plus Mixer Mixing Tubes up to 25 mm Diameter