Learn More
Corning™ Color-Coded Cap Inserts
For use with Corning cryogenic vials except 03-374-6
Brand: Corning™ 2018
Additional Details : Weight : 0.09072kg
Description
- Cap inserts provide color coding for easy sample identification.
- Inserts are packaged in resealable bags.
- Nonsterile
- Cap inserts fit all Corning™ cryogenic vials.
Specifications
Green | |
Polypropylene | |
500 | |
Polypropylene |
Green | |
Non-sterile | |
50 |