Learn More
CD3 Mouse anti-Human, APC, Clone: UCHT1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech APC-65151-100TESTS
Additional Details : Weight : 0.00010kg
Product Code. 16850526
  • 40.00 EUR / 100 tests
Estimated Shipment: 05-06-2024
to see stock.
Description
Description
The CD3 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD3. This antibody recognizes human antigen. The CD3 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
Specifications
CD3
Monoclonal
5 μL (0.25 μg)/Test
Mouse
Primary
Human
Flow Cytometry
UCHT1
APC
100 Tests
916
IgG1