Learn More
Mettler Toledo™ Carrying Case for Densito/DensitoPro
Mettler Toledo Carrying Case for Densito/DensitoPro is ideal carrying the Densito or DensitoPro.
Brand: Mettler Toledo™ 30330861
Additional Details : Weight : 0.90000kg
Description
- Everything you need to perform your measurements fits into the case: your portable density meter, sample tubes, syringes, adapters, etc.
- This durable case protects your instrument out in the field
Specifications
3 kg | |
Accessory | |
Chinese, English (GB), English (US), French, German, Italian, Korean, Polish, Portuguese (BR), Russian, Spanish | |
Use with Densito and DensitoPro Handheld Density Meters |
Carrying Case, Densito | |
20 cm | |
Densito and DensitoPro Handheld Density Meters |