Learn More
Wheaton™ Borosilicate Glass Filtering Flask
Brand: Wheaton™ 635526
293.55 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 1.15200kg
Specifications
2000 mL | |
Borosilicate Glass | |
Joint Connection |