Learn More
Fisherbrand™ Beta Radiation Safe Hourglass Shield
Provides radiation protection shielding for use in laboratories.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
Product Code. 11517253
  • 137.00 EUR / Each
Estimated Shipment: 09-10-2024
Description
  • Transparent shielding
  • Constructed of acrylic provides easy fabrication and manufacturability
Specifications
Specifications
Flat base
Hourglass shield
L x B: 45 x 30 cm (17.71 x 11.81 in.)
Beta radiation protection
Acrylic