missing translation for 'onlineSavingsMsg'
Learn More

ATAGO™ RX-5000α Automatic Digital Refractometer

Can set measurement temperature internally and measures the refractive index, Brix or concentration of various liquids accurately and speedily. The ATAGO™ RX-5000α Automatic Digital Refractometer has a thermo-module to control temperature, so a constant temperature water bath is not necessary.

Brand:  ATAGO™ 3261


Additional Details : Weight : 9.00000kg

Product Code. 10631983

  • 9465.00 EUR / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

  • The measurement starts automatically after the sample reaches your targeted temperature; the refractive index and Brix at your targeted temperature are speedily displayed.
  • High Brix ±0.03% and refractive index ±0.00004 accuracy can be obtained
  • Displays the top-and-bottom-limit-bar for your set control range
  • If measurement value differs from that of your Standard Liquid or other refractometer, it can be adjusted in a certain range
  • 60 kinds of user scales can be input according to your sample
  • Capable of calling up 30 latest measurements
  • Special model of RX-5000α allows pre-programming of an additional scale (such as honey moisture, inverted sugar % or salt concentration) in addition to Refractive Index and Brix (%) at customer request

Services

Two year limited warranty; can be extended to three years if the product is registered with ATAGO.

Specifications

Specifications

Refractometer
1.32700 to 1.58000
Refractive index (nD): ±0.00004; Brix: ±0.03%
Refractive index (nD) : 0.00001; Brix: 0.01%
5°C to 60°C
0.01°C
0.00 to 100.00 Brix
RX-5000α
Refractive index (nD) : ±0.00002; Brix : ±0.01%
Brix, Refractive Index
5°C to 60°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers