Learn More
anti-FUCA1, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand: Abnova H00002517-PW1.1Set
Additional Details : Weight : 0.02000kg
Includes: Antibody pair for IP: rabbit polyclonal anti-FUCA1 (300μL), Antibody pair for WB: mouse purified polyclonal anti-FUCA1 (50μg)
Specifications
FUCA1 | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
FUCA | |
Mouse, Rabbit | |
RUO | |
2517 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
FUCA1 | |
FUCA1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-FUCA1 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-FUCA1 (50 ug) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only