Learn More
Amyloid beta 40 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA316760
Additional Details : Weight : 0.01000kg
Product Code. 11558801
  • 492.00 EUR / 100µL
Estimated Shipment: 30-05-2024
to see stock.
Description
Description
Amyloid beta-protein (abeta) is associated with neuronal injury and death in Alzheimer′s disease. These proteins are the main components of amyloid plaques.
Specifications
Specifications
Amyloid beta 40
Polyclonal
APP
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN2
Rabbit
100 μL
Primary
Human, Mouse
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
P05067, P12023
APP
Synthetic peptide corresponding to amino acid residues G L M V G G VV of human abeta 40.
RUO
11820, 351
-20° C, Avoid Freeze/Thaw Cycles
Liquid