Learn More
Ansell AlphaTec™ 2000 STANDARD Trousers Bound
309.56 EUR valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
X-Large
2X-Large
Description
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Optimized fit, improving wearer comfort and safety
- Low Linting
Specifications
Size | 2X-Large |
Material | Microporous Non-Woven Polyethylene Laminate |
Color | White |
Seam Type | Bound |
Certifications/Compliance (Europe) | Partial Body Protection Only, EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013 |
Style | W20B301 |
For Use With (Application) | Clean Room Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services |