Learn More
Eppendorf™ Adapter for SBS-Format Skirted Plates for Eppendorf™ Rotors A-2-MTP and A-2-DWP
Brand: Eppendorf™ 5825718003
152.00 EUR valid until 2024-12-31
Use promo code "23292" to get your promotional price.
Additional Details : Weight : 0.08000kg
Specifications
Adapter | |
Rotors A-2-MTP and A-2-DWP | |
230 V |