missing translation for 'onlineSavingsMsg'
Learn More

ADAM10 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA587899

Additional Details : Weight : 0.01000kg

Product Code. 16546261

  • 470.00 EUR / 100µL
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Positive Samples: HT-1080, Raji, Jurkat, Mouse kidney, Mouse brain; Cellular Location: Cell membrane, Endomembrane system, Single-pass type I membrane protein Immunogen sequence: EKNTCQLYIQ TDHLFFKYYG TREAVIAQIS SHVKAIDTIY QTTDFSGIRN ISFMVKRIRI NTTADEKDPT NPFRFPNIGV EKFLELNSEQ NHDDYCLAYV FTDRDFDDGV LGLAWVGAPS GSSGGICEKS KLYSDGKKKS LNTGIITVQN YGSHVPPKVS HITFAHEVGH NFGSPHDSGT ECTPGESKNL GQKENGNYIM YARATSGDKL NNNKFS

Proinflammatory cytokine tumor necrosis factor-a (TNF-a) contributes to a variety of inflammatory responses and programmed cell death. Notch receptor and its ligand participate in cell fate decisions during vertebrate development and are associated with several human disorders, including a T-cell lymphoma. TNF-a, notch and its ligand delta are all membrane-bond molecules, which are cleaved by proteases to release mature proteins or functional receptor. ADAM10, a metalloprotease-disintegrin in the family of mammalian ADAM (for a disintegrin and metalloprotease), was recently identified to cleave TNF-a, notch and its ligand delta (1-3). The genes encoding human, mouse, and bovine ADAM10 were recently cloned and designated ADAM 10, kuzbanian (KUZ), and MADM, respectively. ADAM10 mRNA is expressed in a variety of human and bovine tissues.
Specifications

Specifications

ADAM10
Polyclonal
Unconjugated
Adam10
1700031C13Rik, a disintegrin and metallopeptidase domain 10, a disintegrin and metalloprotease domain (ADAM) 10, a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase, AD10, AD18, ADAM, ADAM 10, ADAM metallopeptidase domain 10, ADAM10, ADAMs, CD156c, CDw156, Disintegrin and metalloproteinase domain-containing protein 10, HsT18717, KUZ, kuzbanian, kuzbanian protein homolog, MADM, Mammalian disintegrin-metalloprotease, metalloendopeptidases, metalloproteinase, myelin-associated metalloproteinase, RAK
Rabbit
Affinity Chromatography
RUO
102, 11487
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Western Blot
0.62 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300, pH 7.3
O14672, O35598
Adam10
Recombinant fusion protein containing a sequence corresponding to amino acids 218-433 of human ADAM10 (NP_0011011)
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers