missing translation for 'onlineSavingsMsg'
Learn More

Acipimox, Thermo Scientific™

Catalog Number 17555535
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
51037-30-0
C6 H6 N2 O3
This item is not returnable. View return policy
51037-30-0
C6 H6 N2 O3
Quantity 5 g
Solubility Soluble in methanol
Merck Index 13-113
Melting Point 180°C to 190°C
Formula Weight 154.12
Physical Form Powder
Color White to Almost-white
Purity ≥98.0%
InChI Key DNRXJHATQULEHC-UHFFFAOYSA-N
SMILES CC1=CN=CC(C(O)=O)=[N+]1[O-]
IUPAC Name 2-carboxy-6-methylpyrazin-1-ium-1-olate
Molecular Weight (g/mol) 154.13
Chemical Name or Material Acipimox
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'