Learn More
Viking Optical™ 5X Folding Magnifier
Brand:  Viking Optical™ 5607
Additional Details : Weight : 0.01260kg
Product Code. 12866325
  • 3.04 EUR / Each
Estimated Shipment: 25-06-2024
Description
Pocket doublet type, giving tolerably flat field with minimum chromatic aberration. Lens approximately 19mm diameter. Mounted in metal frame.
Specifications
Specifications
Metal Frame
19 mm
Folding Magnifier
5X