missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Specifications
CAS | 269410-24-4 |
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Molecular Formula | C14H18BNO2 |
MDL Number | MFCD03789263 |
Quantity | 1 g |
Synonym | 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 5-indoleboronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, indole-5-boronic acid pinacol ester, 1h-indole-5-boronic acid pinacol ester, 5-indolylboronic acid, pinacol ester, 1h-indole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, indolyl-5-boronic acid pinacol ester, indole-5-boronic acid, pinacol ester |
InChI Key | FATPQDPUKVVCLT-UHFFFAOYSA-N |
SMILES | CC1(C)OB(OC1(C)C)C1=CC=C2NC=CC2=C1 |
IUPAC Name | 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'