missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzonitrile, 98%

Product Code. p-7028001
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406137 5 g 5g
11416137 25 g 25g
11426137 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11406137 Supplier Thermo Scientific Alfa Aesar Supplier No. B25104.06
Add to basket
This item is not returnable. View return policy

CAS: 3058-39-7 | C7H4IN | 229.02 g/mol

4-Iodobenzonitrile is used in organic synthesis. It is used as catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzonitrile is used in organic synthesis. It is used as catalytic agent, petrochemical additive.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in the dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3058-39-7
Molecular Formula C7H4IN
Molecular Weight (g/mol) 229.02
MDL Number MFCD00051310
InChI Key XOKDXPVXJWTSRM-UHFFFAOYSA-N
Synonym p-iodobenzonitrile, benzonitrile, 4-iodo, p-cyanoiodobenzene, benzonitrile, p-iodo, 1-cyano-4-iodobenzene, 4-cyanoiodobenzene, 4-iodo-benzonitrile, 4-iodobenzenecarbonitrile, 4-iodbenzonitril, 4-iodo benzonitrile
PubChem CID 76467
IUPAC Name 4-iodobenzonitrile
SMILES IC1=CC=C(C=C1)C#N

Specifications

Melting Point 125°C to 127°C
Quantity 5 g
UN Number UN3439
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 229.02
Percent Purity 98%
Chemical Name or Material 4-Iodobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.