missing translation for 'onlineSavingsMsg'
Learn More

3'-Nitroacetophenone, 98+%

Product Code. p-7081930
Click to view available options
Quantity:
250 g
500 g
1000 g
Unit Size:
1000g
250g
500g
This item is not returnable. View return policy

Product Code. 11440133

Brand: Thermo Scientific Alfa Aesar A12155.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121-89-1 | C8H7NO3 | 165.148 g/mol

3'-Nitroacetophenone is used as an intermediate in pharmaceutical industry for making drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Nitroacetophenone is used as an intermediate in pharmaceutical industry for making drugs.

Solubility
Soluble in water 0.75 g/L @ 20°C.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-89-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.148
MDL Number MFCD00007259
InChI Key ARKIFHPFTHVKDT-UHFFFAOYSA-N
Synonym 3'-nitroacetophenone, 3-nitroacetophenone, m-nitroacetophenone, 1-3-nitrophenyl ethanone, ethanone, 1-3-nitrophenyl, acetophenone, 3'-nitro, m-acetylnitrobenzene, usaf ma-1, methyl 3-nitrophenyl ketone, 3-nitroacetofenon
PubChem CID 8494
IUPAC Name 1-(3-nitrophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 76°C to 80°C
Boiling Point 202°C
Odor Slightly sweetish
Quantity 250 g
Beilstein 743002
Solubility Information Soluble in water 0.75g/L @ 20°C.
Formula Weight 165.15
Percent Purity ≥98%
Chemical Name or Material 3'-Nitroacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.